Merck
  • Synthesis, structural elucidation, DNA-PK inhibition, homology modelling and anti-platelet activity of morpholino-substituted-1,3-naphth-oxazines.
Synthesis, structural elucidation, DNA-PK inhibition, homology modelling and anti-platelet activity of morpholino-substituted-1,3-naphth-oxazines.
Bioorganic & medicinal chemistry (2011-06-15)
Saleh Ihmaid, Jasim Al-Rawi, Christopher Bradley, Michael J Angove, Murray N Robertson, Rachel L Clark
ABSTRACT
A number of new angular 2-morpholino-(substituted)-naphth-1,3-oxazines (compound 10b), linear 2-morpholino-(substituted)-naphth-1,3-oxazines (compounds 13b-c), linear 6, 7 and 9-O-substituted-2-morpholino-(substituted)-naphth-1,3-oxazines (compounds 17-22, 24, and 25) and angular compounds 14-16 and 23 were synthesised. The O-substituent was pyridin-2yl-methyl (15, 18, and 21) pyridin-3yl-methyl (16, 19, and 22) and 4-methylpipreazin-1-yl-ethoxy (23-25). Twelve compounds were tested for their inhibitory effect on collagen induced platelet aggregation and it was found that the most active compounds were compounds 19 and 22 with IC(50)=55±4 and 85±4 μM, respectively. Furthermore, the compounds were also assayed for their ability to inhibit DNA-dependent protein kinase (DNA-PK) activity. The most active compounds were 18 IC(50)=0.091 μM, 24 IC(50)=0.191 μM, and 22 IC(50)=0.331 μM. Homology modelling was used to build a 3D model of DNA-PK based on the X-ray structure of phosphatidylinositol 3-kinases (PI3Ks). Docking of synthesised compounds within the binding pocket and structure-activity relationships (SAR) analyses of the poses were performed and results agreed well with observed activity.
Sigma-Aldrich
Morpholine, ACS reagent, ≥99.0%
Supelco
Morpholine, analytical standard
Sigma-Aldrich
Morpholine, ReagentPlus®, ≥99%
Sigma-Aldrich
Morpholine, purified by redistillation, ≥99.5%