Merck
  • Detections of eleven organophosphorus insecticides and one herbicide threatening Pacific salmonids, Oncorhynchus spp., in California, 1991-2010.
Detections of eleven organophosphorus insecticides and one herbicide threatening Pacific salmonids, Oncorhynchus spp., in California, 1991-2010.
Bulletin of environmental contamination and toxicology (2011-06-29)
Emily B Lisker, Michael P Ensminger, Sheryl L Gill, Kean S Goh
ABSTRACT
California's surface water monitoring results from 1991 through 2010 were analyzed to determine whether 12 organophosphorus insecticides and herbicides (i.e., azinphos methyl, bensulide, dimethoate, disulfoton, ethoprop, fenamiphos, methamidophos, methidathion, methyl parathion, naled, phorate, and phosmet) and their degradates have been detected above maximum concentration limits (MCLs) in Pacific salmonid habitats. Methidathion, methyl parathion, phorate, phosmet, and the oxygen analogue of naled (DDVP) detections exceeded MCLs. Methyl parathion detections may be accounted for by monthly use trends, while methidathion detections may be explained by yearly use trends. There were inadequate phorate, phosmet, or DDVP data to evaluate for correlations with use.
Product Description
Supelco
Disulfoton, PESTANAL®, analytical standard
Supelco
Ethoprophos, PESTANAL®, analytical standard
Supelco
Phorate, PESTANAL®, analytical standard
Supelco
Methidathion, PESTANAL®, analytical standard
Supelco
Methamidophos, PESTANAL®, analytical standard
Supelco
Fenamiphos, PESTANAL®, analytical standard