Merck
  • Synthesis, crystal structure and luminescent properties of one 3D Cd(II) coordination polymer [Cd(H3BPTC)2(bpy)]n (H4BPTC = 1,1'-biphenyl-2,2',6,6'-tetracarboxylic acid, bpy = 4,4'-bipyridine).
Synthesis, crystal structure and luminescent properties of one 3D Cd(II) coordination polymer [Cd(H3BPTC)2(bpy)]n (H4BPTC = 1,1'-biphenyl-2,2',6,6'-tetracarboxylic acid, bpy = 4,4'-bipyridine).
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2011-07-29)
Chong-Zhen Mei, Wen-Wen Shan, Bing-Tao Liu
ABSTRACT
A new 3D metal-organic coordination polymer [Cd(H(3)BPTC)(2)(bpy)](n) (1) (H(4)BPTC = 1,1'-biphenyl-2,2',6,6'-tetracarboxylic acid, bpy =4,4'-bipyridine) has been synthesized and characterized by single X-ray diffraction and IR spectroscopy. The one-dimensional metal-organic chains of the title complex, namely [Cd(H(3)BPTC)(2)](n), are held together through hydrogen bonding and bridging "second" ligand 4,4'-bpy to give a three-dimensional metal-organic network. The thermal stability of complex 1 was studied by thermal gravimetric (TG) and differential thermal analysis (DTA). Compound 1 exhibits photoluminescence with an emission maximum at ca. 380 nm upon excitation at ca. 251 nm.
Sigma-Aldrich
4,4′−Dipyridyl, 98%