Merck
Hydrogenated cation vacancies in semiconducting oxides.
Journal of physics. Condensed matter : an Institute of Physics journal (2011-08-05)
J B Varley, H Peelaers, A Janotti, C G Van de Walle
ABSTRACT
Using first-principles calculations we have studied the electronic and structural properties of cation vacancies and their complexes with hydrogen impurities in SnO(2), In(2)O(3) and β-Ga(2)O(3). We find that cation vacancies have high formation energies in SnO(2) and In(2)O(3) even in the most favorable conditions. Their formation energies are significantly lower in β-Ga(2)O(3). Cation vacancies, which are compensating acceptors, strongly interact with H impurities resulting in complexes with low formation energies and large binding energies, stable up to temperatures over 730 °C. Our results indicate that hydrogen has beneficial effects on the conductivity of transparent conducting oxides: it increases the carrier concentration by acting as a donor in the form of isolated interstitials, and by passivating compensating acceptors such as cation vacancies; in addition, it potentially enhances carrier mobility by reducing the charge of negatively charged scattering centers. We have also computed vibrational frequencies associated with the isolated and complexed hydrogen, to aid in the microscopic identification of centers observed by vibrational spectroscopy.
Sigma-Aldrich
Gallium(III) oxide, ≥99.99% trace metals basis
Sigma-Aldrich
Indium(III) oxide, 99.998% trace metals basis
Sigma-Aldrich
Indium(III) oxide, 99.99% trace metals basis
Sigma-Aldrich
Indium(III) oxide, nanopowder, <100 nm particle size (TEM), 99.9% trace metals basis