Merck
  • Structural insight into the stereoselective production of PGF2α by Old Yellow Enzyme from Trypanosoma cruzi.
Structural insight into the stereoselective production of PGF2α by Old Yellow Enzyme from Trypanosoma cruzi.
Journal of biochemistry (2011-08-16)
Naoki Okamoto, Keishi Yamaguchi, Eiichi Mizohata, Keiji Tokuoka, Nahoko Uchiyama, Shigeru Sugiyama, Hiroyoshi Matsumura, Koji Inaka, Yoshihiro Urade, Tsuyoshi Inoue
ABSTRACT
Old yellow enzyme (OYE) is an NADPH oxidoreductase capable of reducing a variety of compounds. It contains flavin mononucleotide (FMN) as a prosthetic group. A ternary complex structure of OYE from Trypanosoma cruzi (TcOYE) with FMN and one of the substrates, p-hydroxybenzaldehyde, shows a striking movement around the active site upon binding of the substrate. From a structural comparison of other OYE complexed with 12-oxophytodienoate, we have constructed a complex structure with another substrate, prostaglandin H(2) (PGH(2)), to provide a proposed stereoselective reaction mechanism for the reduction of PGH(2) to prostaglandin F(2α) by TcOYE.
Sigma-Aldrich
4-Hydroxybenzaldehyde, ≥97%, FG
Sigma-Aldrich
4-Hydroxybenzaldehyde, 98%
Sigma-Aldrich
4-Hydroxybenzaldehyde, ≥95.0% (HPLC)