Merck
  • Oxidative properties of a nonheme Ni(II)(O2) complex: Reactivity patterns for C-H activation, aromatic hydroxylation and heteroatom oxidation.
Oxidative properties of a nonheme Ni(II)(O2) complex: Reactivity patterns for C-H activation, aromatic hydroxylation and heteroatom oxidation.
Chemical communications (Cambridge, England) (2011-09-06)
Reza Latifi, Laleh Tahsini, Devesh Kumar, G Narahari Sastry, Wonwoo Nam, Sam P de Visser
ABSTRACT
Density functional theory calculations on the reactivity of a Ni(II)-superoxo complex in C-H bond activation, aromatic hydroxylation and heteroatom oxidation reactions have been explored; the Ni(II)-superoxo complex is able to react with substrates with weak C-H bonds and PPh(3).
Sigma-Aldrich
1,4-Cyclohexadiene, purum, ≥97.0% (GC)
Sigma-Aldrich
1,4-Cyclohexadiene, 97%