Merck
  • Application of headspace solid phase dynamic extraction gas chromatography/mass spectrometry (HS-SPDE-GC/MS) for biomonitoring of n-heptane and its metabolites in blood.
Application of headspace solid phase dynamic extraction gas chromatography/mass spectrometry (HS-SPDE-GC/MS) for biomonitoring of n-heptane and its metabolites in blood.
Toxicology letters (2011-09-10)
Bernd Rossbach, Peter Kegel, Stephan Letzel
ABSTRACT
Solid phase dynamic extraction (SPDE) is an innovative sample preparation and enrichment technique in connection with gas chromatography (GC). Using SPDE, we developed a method for simultaneous determination of n-heptane and its mono-oxygenated metabolites heptane-4-one, 3-one, 2-one, 4-ol, 3-ol, 2-ol, and 1-ol in blood. After adjustment of various extraction and desorption parameters, method validation resulted in limits of detection (LOD) between 0.006 (heptane-4-one) and 0.021mg/L (heptane-1-ol). Intra-assay coefficients of variation ranged between 4.8% and 20.8% while relative recovery ranged between 100% and 117% (spiked concentration 0.128mg/L, n=8). The method was applied to blood samples, which have been collected from 20 volunteers after controlled inhalative exposure to 167, 333, and 500ppm n-heptane. After 3h of exposure, n-heptane and heptane-2-one were detectable in all samples in concentrations ranging up to 2.903 and 0.495mg/L, while the concentrations of the remaining analytes were closer to the respective LOD or even below. A significant linear relationship with ambient exposure (R(2)=0.701, p<0.001, n=55) was found for n-heptane in blood, which could be helpful for evaluation of biological exposure limits in future. Due to its high abundance in blood, 2-heptanone could be an interesting candidate as a biomarker also in alternative matrices such as urine or saliva.
Sigma-Aldrich
Heptane, anhydrous, 99%
Sigma-Aldrich
Heptane, biotech. grade, ≥99%
Supelco
Heptane, analytical standard
Sigma-Aldrich
Heptane, puriss., ≥99% (GC)
Sigma-Aldrich
Heptane, ReagentPlus®, 99%
Sigma-Aldrich
Heptane, suitable for HPLC, ≥99%
Sigma-Aldrich
Heptane, suitable for HPLC, ≥96%
Sigma-Aldrich
Heptane, HPLC Plus, for HPLC, GC, and residue analysis, 99%