Merck
  • A review of the receptor-binding properties of p-synephrine as related to its pharmacological effects.
A review of the receptor-binding properties of p-synephrine as related to its pharmacological effects.
Oxidative medicine and cellular longevity (2011-09-10)
Sidney J Stohs, Harry G Preuss, Mohd Shara
ABSTRACT
Bitter orange (Citrus aurantium) extract and its primary protoalkaloid p-synephrine are used widely in weight loss/weight management and sports performance products. Because of structural similarities, the pharmacological effects of p-synephrine are widely assumed to be similar to those of ephedrine, m-synephrine (phenylephrine), and endogenous amine neurotransmitters as norepinephrine and epinephrine. However, small structural changes result in the receptor binding characteristics of these amines that are markedly different, providing a plausible explanation for the paucity of adverse effects associated with the wide-spread consumption of p-synephrine in the form of dietary supplements as well as in various Citrus foods and juices. This paper summarizes the adrenoreceptor binding characteristics of p-synephrine relative to m-synephrine, norepinephrine, and other amines as related to the observed pharmacological effects.