Merck
  • Isolation and functional assessment of mitochondria from small amounts of mouse brain tissue.
Isolation and functional assessment of mitochondria from small amounts of mouse brain tissue.
Methods in molecular biology (Clifton, N.J.) (2011-09-14)
Christos Chinopoulos, Steven F Zhang, Bobby Thomas, Vadim Ten, Anatoly A Starkov
ABSTRACT
Recent discoveries have brought mitochondria functions in focus of the neuroscience research community and greatly stimulated the demand for approaches to study mitochondria dysfunction in neurodegenerative diseases. Many mouse disease models have been generated, but studying mitochondria isolated from individual mouse brain regions is a challenge because of small amount of the available brain tissue. Conventional techniques for isolation and purification of mitochondria from mouse brain subregions, such as ventral midbrain, hippocampus, or striatum, require pooling brain tissue from six to nine animals for a single mitochondrial preparation. Working with pooled tissue significantly decreases the quality of data because of the time required to dissect several brains. It also greatly increases the labor intensity and the cost of experiments as several animals are required per single data point. We describe a method for isolation of brain mitochondria from mouse striata or other 7-12 mg brain samples. The method utilizes a refrigerated table-top microtube centrifuge, and produces research grade quality mitochondria in amounts sufficient for performing multiple enzymatic and functional assays, thereby eliminating the necessity for pooling mouse brain tissue. We also include a method of measuring ADP-ATP exchange rate as a function of mitochondrial membrane potential (ΔΨm) in small amounts of isolated mitochondria, adapted to a plate reader format.
Sigma-Aldrich
Safranin O, Dye content ≥85 %
Sigma-Aldrich
Safranin O, certified by the BSC
Sigma-Aldrich
Safranin O, suitable for microscopy
Sigma-Aldrich
Schaeffer and Fulton Spore Stain Solution B
Sigma-Aldrich
Gram′s safranin solution, suitable for microscopy