Merck
  • Upregulation of cardiomyocyte ribonucleotide reductase increases intracellular 2 deoxy-ATP, contractility, and relaxation.
Upregulation of cardiomyocyte ribonucleotide reductase increases intracellular 2 deoxy-ATP, contractility, and relaxation.
Journal of molecular and cellular cardiology (2011-09-20)
F S Korte, Jin Dai, Kate Buckley, Erik R Feest, Nancy Adamek, Michael A Geeves, Charles E Murry, Michael Regnier
ABSTRACT
We have previously demonstrated that substitution of ATP with 2 deoxy-ATP (dATP) increased the magnitude and rate of force production at all levels of Ca(2+)-mediated activation in demembranated cardiac muscle. In the current study we hypothesized that cellular [dATP] could be increased by viral-mediated overexpression of the ribonucleotide reductase (Rrm1 and Rrm2) complex, which would increase contractility of adult rat cardiomyocytes. Cell length and ratiometric (Fura2) Ca(2+) fluorescence were monitored by video microscopy. At 0.5Hz stimulation, the extent of shortening was increased ~40% and maximal rate of shortening was increased ~80% in cardiomyocytes overexpressing Rrm1+Rrm2 as compared to non-transduced cardiomyocytes. The maximal rate of relaxation was also increased ~150% with Rrm1+Rrm2 overexpression, resulting in decreased time to 50% relaxation over non-transduced cardiomyocytes. These differences were even more dramatic when compared to cardiomyocytes expressing GFP-only. Interestingly, Rrm1+Rrm2 overexpression had no effect on minimal or maximal intracellular [Ca(2+)], indicating increased contractility is primarily due to increased myofilament activity without altering Ca(2+) release from the sarcoplasmic reticulum. Additionally, functional potentiation was maintained with Rrm1+Rrm2 overexpression as stimulation frequency was increased (1Hz and 2Hz). HPLC analysis indicated cellular [dATP] was increased by approximately 10-fold following transduction, becoming ~1.5% of the adenine nucleotide pool. Furthermore, 2% dATP was sufficient to significantly increase crossbridge binding and contractile force during sub-maximal Ca(2+) activation in demembranated cardiac muscle. These experiments demonstrate the feasibility of directly targeting the actin-myosin chemomechanical crossbridge cycle to enhance cardiac contractility and relaxation without affecting minimal or maximal Ca(2+). This article is part of a Special issue entitled "Possible Editorial".
Sigma-Aldrich
2′-Deoxyadenosine 5′-triphosphate sodium salt solution, 10 mM
Sigma-Aldrich
2′-Deoxyadenosine 5′-triphosphate sodium salt solution, 100 mM, pH 7
Sigma-Aldrich
2′-Deoxyadenosine 5′-triphosphate disodium salt, ≥97%