Merck
  • Dicationic ion-pairing of phosphoric acid diesters post-liquid chromatography and subsequent determination by electrospray positive ionization-tandem mass spectrometry.
Dicationic ion-pairing of phosphoric acid diesters post-liquid chromatography and subsequent determination by electrospray positive ionization-tandem mass spectrometry.
Journal of chromatography. A (2011-09-29)
Shaogang Chu, Da Chen, Robert J Letcher
ABSTRACT
Several organophosphate triesters are widely used as flame retardants and can be metabolized to dibutyl (DBP), diphenyl (DPhP), di(2-ethylhexyl) (DEHP) and di(1,3-dichloro-2-propyl) (or bis(1,3-dichloro-2-propyl); DDCPP) phosphoric acid, respectively. A highly sensitive liquid chromatography-electrospray ionization(+)-triple quadrupole mass spectrometry (LC-ESI(+)-QQQ-MS/MS) based analysis method was presently developed. In this method the target compounds were separated with a C(18)-based reversed phase LC column, and decamethonium hydroxide (dicatonic reagent) was introduced post-LC to form ion-pairs, which were subsequently detected by ESI(+). For the phosphate acid diester ion-pairs, the mass spectra showed the most abundant ion to be [(CH(3))(2)N(CH(2))(10)N(CH(3))(3)](+), with lesser abundances of [[M-H](-)[(CH(3))(3)N(CH(2))(9)CH(2)](2+)](+) and [CH(2)CH(CH(2))(8)N(CH(3))(3)](+). For DDCPP, the fragment ions of [[Cl](-)[(CH(3))(3)N(CH(2))(10)N(CH(3))(3)](2+)](+) and [[Cl](-)[(CH(3))(3)N(CH(2))(9)CH(2)](2+)](+) could also be observed. The limits of quantitation (LOQs) by LC-ESI(+)-MS/MS (based on multiple reaction monitoring) were 0.14, 0.03, 0.14 and 0.02 ng/mL for DPhP, DBP, DDCPP and DEHP, respectively. The response was highly linearly correlated (r>0.995) with concentration over the range of the LOD to 1000 ng/mL. The matrix effect on ESI+ was negligible for the samples in experiment of in vitro metabolism using rat liver microsomes.
Sigma-Aldrich
Decamethonium bromide, Ion-Pair reagent, ≥99%
Sigma-Aldrich
Decamethonium bromide, crystalline