Merck
  • Indigenous soil bacteria with the combined potential for hydrocarbon consumption and heavy metal resistance.
Indigenous soil bacteria with the combined potential for hydrocarbon consumption and heavy metal resistance.
Environmental science and pollution research international (2011-09-29)
Nida Ali, Narjes Dashti, Dina Al-Mailem, Mohamed Eliyas, Samir Radwan
ABSTRACT
Transconjugant bacteria with combined potential for hydrocarbon utilization and heavy metal resistance were suggested by earlier investigators for bioremediation of soils co-contaminated with hydrocarbons and heavy metals. The purpose of this study was to offer evidence that such microorganisms are already part of the indigenous soil microflora. Microorganisms in pristine and oily soils were counted on nutrient agar and a mineral medium with oil as a sole carbon source, in the absence and presence of either sodium arsenate (As V), sodium arsenite (As III) or cadmium sulfate, and characterized via 16S rRNA gene sequencing. The hydrocarbon-consumption potential of individual strains in the presence and absence of heavy metal salts was measured. Pristine and oil-contaminated soil samples harbored indigenous bacteria with the combined potential for hydrocarbon utilization and As and Cd resistance in numbers up to 4 × 10⁵ CFU g⁻¹. Unicellular bacteria were affiliated to the following species arranged in decreasing order of predominance: Bacillus subtilis, Corynebacterium pseudotuberculosis, Brevibacterium linens, Alcaligenes faecalis, Enterobacter aerogenes, and Chromobacterium orangum. Filamentous forms were affiliated to Nocardia corallina, Streptomyces flavovirens, Micromonospora chalcea, and Nocardia paraffinea. All these isolates could grow on a wide range of pure aliphatic and aromatic hydrocarbons, as sole sources of carbon and energy, and could consume oil and pure hydrocarbons in batch cultures. Low As concentrations, and to a lesser extent Cd concentrations, enhanced the hydrocarbon-consumption potential by the individual isolates. There is no need for molecularly designing microorganisms with the combined potential for hydrocarbon utilization and heavy metal resistance, because they are already a part of the indigenous soil microflora.
Sigma-Aldrich
Sodium arsenate dibasic heptahydrate, ≥98.0%
Sigma-Aldrich
Sodium arsenate dibasic heptahydrate, ACS reagent, ≥98%
Sigma-Aldrich
Octadecane, 99%
Supelco
Octadecane, analytical standard
Sigma-Aldrich
Cadmium sulfate 8/3-hydrate, puriss. p.a., ACS reagent, ≥99.0% (calc. based on CdSO4 · 8/3 H2O, KT)
Sigma-Aldrich
Cadmium sulfate, ACS reagent, ≥99.0%