Merck
  • Effects of cytokinins, cytokinin ribosides and their analogs on the viability of normal and neoplastic human cells.
Effects of cytokinins, cytokinin ribosides and their analogs on the viability of normal and neoplastic human cells.
Anticancer research (2011-10-04)
Silvana Casati, Roberta Ottria, Erika Baldoli, Elisa Lopez, Jeanette A M Maier, Pierangela Ciuffreda
ABSTRACT
We examined the effects of some cytokinins and cytokinin ribosides including a series of adenosine analogs differently substituted in the N(6) position, along with some hypoxanthine derivatives on the viability of normal and neoplastic human cells. Cytokinins such as trans-zeatin, isopentenyladenine and benzyladenine do not show any effect, while cytokinin ribosides such as trans-zeatin riboside, isopentenyladenosine, and benzylaminopurine riboside impair the viability of normal and neoplastic cells, apart from colon carcinoma LoVo cells.
Sigma-Aldrich
trans-Zeatin-riboside, ~95%
Sigma-Aldrich
trans-Zeatin-riboside, suitable for plant cell culture, BioReagent, ~95%
Sigma-Aldrich
trans-Zeatin, suitable for plant cell culture, BioReagent, ≥97%