Merck
  • Adsorption of four perfluorinated acids on non ion exchange polymer sorbents.
Adsorption of four perfluorinated acids on non ion exchange polymer sorbents.
Water science and technology : a journal of the International Association on Water Pollution Research (2011-10-08)
S T M L D Senevirathna, S Tanaka, S Fujii, C Kunacheva, H Harada, B R Shivakoti, H Dinh, T Ariyadasa
ABSTRACT
Perfluorinated compounds (PFCs) have attracted global concern due to their ubiquitous distribution and properties of persistence, bio accumulation and toxicity. The process of adsorption has been identified as an effective technique to remove PFCs in water. Different non ion-exchange polymeric adsorbents were tested with regard to their sorption kinetics and isotherms at low PFCs concentrations. Selected PFCs were perfluorobutanoic acid (PFBA), perfluoroheptanoic acid (PFHpA), perfluorooctanoic acid (PFOA) and perfluorodecanoic acid (PFDA) and the tested polymers were three types of Dowex optopores (V-493, V503, and L493), Amberlite XAD-4, and Filtrasorb 400 (Granular Activated Carbon-GAC). We observed the selective adsorption of PFCs on synthetic polymers. For PFDA, Amberlite XAD-4 gave the Freundlich adsorption constant of 2,965 (microg PFCs/g sorbent)(microg PFCs/L)(-n), which was higher than that of GAC (121.89 (microg PFCs/g sorbent) (microg PFCS/L)(-n)). In the case of PFBA, GAC showed better performance (13.36) (microg PFCs/g sorbent) microg PFCS/L)(-n) than synthetic polymers (0.62-5.23) (microg PFCs/g sorbent) (microg PFCS/L)(-n). Adsorption kinetics of all adsorbents were well described (R2 = 0.85-1) by pseudo-second order kinetic model. Sorption capacity was influenced by initial PFCs concentration for all adsorbents. GAC reached the equilibrium concentration within 4 hours, Amberlite XAD 4 reached it within 10 hours and other polymers took more than 70 hours.
Product Description
Supelco
Heptafluorobutyric acid solution, 0.5 M in H2O, LiChropur, suitable for ion pair chromatography
Sigma-Aldrich
Heptafluorobutyric acid, ≥99.0% (GC)
Sigma-Aldrich
Heptafluorobutyric acid, 98%
Supelco
Heptafluorobutyric acid, ≥99.5% (GC)
Sigma-Aldrich
Perfluorodecanoic acid, 97%