Merck
  • Dihydrogen addition in a dinuclear rare-earth metal hydride complex supported by a metalated TREN ligand.
Dihydrogen addition in a dinuclear rare-earth metal hydride complex supported by a metalated TREN ligand.
Journal of the American Chemical Society (2011-10-12)
Ajay Venugopal, Waldemar Fegler, Thomas P Spaniol, Laurent Maron, Jun Okuda
ABSTRACT
The dinuclear lutetium dihydride dication supported by metalated tripodal ligands undergoes facile hydrogenolysis with H(2) to form a trihydride dication. Molecular orbital analysis shows that the LUMO is a bonding Lu···Lu orbital that is poised to activate dihydrogen.
Sigma-Aldrich
Tris(2-aminoethyl)amine, 96%