Merck
Ligand field-tuned single-molecule magnet behaviour of 2p-4f complexes.
Dalton transactions (Cambridge, England : 2003) (2011-11-02)
Xue-Lan Mei, Yue Ma, Li-Cun Li, Dai-Zheng Liao
ABSTRACT
Three new 2p-4f complexes of [Ln(acac)(3)(NIT-2Py)]·0.5NIT-2Py [Ln(III) = Gd(1), Dy(2)] and [Dy(tfa)(3)(NIT-2Py)]·0.5C(7)H(16) (3) (NIT-2Py = 2-(2'-pyridyl)- 4,4,5,5-tetramethylimidazoline-1-oxyl-3-oxide; acac = acetylacetonate and tfa = trifluoroacetylacetonate) have been synthesized, and structurally and magnetically characterized. The X-ray structural analysis exhibits that the three complexes show similar mononuclear structures, in which NIT-2Py radical chelates the Ln(III) ion through the oxygen atom of the NO group and the nitrogen atom from the pyridine ring. The static magnetic measurements on the three complexes exhibit ferromagnetic coupling between the lanthanide ion and the radical. Compared to the silence of the out-of-phase ac susceptibility of complex 3, the magnetic relaxation behavior of complex 2 is observed, suggesting single-molecule magnet behavior. The different magnetic relaxation behaviours of 2 and 3 are due to their slightly different crystal structure around the Dy(III) ions. It was demonstrated that the spin dynamic can be modified by the careful adjustment of the ligand field around the metal center.
Sigma-Aldrich
Copper(II) acetylacetonate, ≥99.9% trace metals basis
Sigma-Aldrich
Copper(II) acetylacetonate, 97%
Sigma-Aldrich
Manganese(III) acetylacetonate, technical grade
Sigma-Aldrich
Dysprosium, ingot, 99.9% trace rare earth metals basis
Sigma-Aldrich
Chromium(III) acetylacetonate, 97%
Sigma-Aldrich
Dysprosium, chips, 99.9% trace rare earth metals basis
Sigma-Aldrich
Manganese(II) acetylacetonate