Merck
Relation between mitochondrial membrane potential and ROS formation.
Methods in molecular biology (Clifton, N.J.) (2011-11-08)
Jan M Suski, Magdalena Lebiedzinska, Massimo Bonora, Paolo Pinton, Jerzy Duszynski, Mariusz R Wieckowski
ABSTRACT
Mitochondria are considered as the main source of reactive oxygen species (ROS) in the cell. For this reason, they have been recognized as a source of various pathological conditions as well as aging. Chronic increase in the rate of ROS production is responsible for the accumulation of ROS-associated damages in DNA, proteins, and lipids, and may result in progressive cell dysfunctions and, in a consequence, apoptosis, increasing the overall probability of an organism's pathological conditions. The superoxide anion is the main undesired by-product of mitochondrial oxidative phosphorylation. Its production is triggered by a leak of electrons from the mitochondrial respiratory chain and the reaction of these electrons with O(2). Superoxide dismutase (MnSOD, SOD2) from the mitochondrial matrix as well as superoxide dismutase (Cu/ZnSOD, SOD1) present in small amounts in the mitochondrial intramembrane space, convert superoxide anion to hydrogen peroxide, which can be then converted by catalase to harmless H(2)O. In this chapter, we describe a relation between mitochondrial membrane potential and the rate of ROS formation. We present different methods applicable for isolated mitochondria or intact cells. We also present experiments demonstrating that a magnitude and a direction (increase or decrease) of a change in mitochondrial ROS production depends on the metabolic state of this organelle.
Sigma-Aldrich
Schaeffer and Fulton Spore Stain Solution B
Pricing and availability is not currently available.
Sigma-Aldrich
Gram′s safranin solution, suitable for microscopy
Pricing and availability is not currently available.
Sigma-Aldrich
Safranin O, suitable for microscopy
Pricing and availability is not currently available.
Sigma-Aldrich
Safranin O, Dye content ≥85 %
Pricing and availability is not currently available.
Sigma-Aldrich
Safranin O, certified by the BSC
Pricing and availability is not currently available.