Merck
  • Effect of antioxidant mixtures on growth and ochratoxin a production of Aspergillus section Nigri species under different water activity conditions on peanut meal extract agar.
Effect of antioxidant mixtures on growth and ochratoxin a production of Aspergillus section Nigri species under different water activity conditions on peanut meal extract agar.
Toxins (2010-06-01)
Carla Barberis, Andrea Astoreca, María Guillermina Fernandez-Juri, Ana María Dalcero, Carina Magnoli
ABSTRACT
The effect of mixtures of antioxidants butylated hydroxyanisol (BHA) and propyl paraben (PP) on lag phase, growth rate and ochratoxin A (OTA) production by four Aspergillus section Nigri strains was evaluated on peanut meal extract agar (PMEA) under different water activities (a(w)). The antioxidant mixtures used were: BHA + PP (mM), M1 (0.5 + 0.5), M2 (1.0 + 0.5), M3 (2.5 + 0.5), M4 (0.5 + 1.0), M5 (1.0 + 1.0), M6 (2.5 + 1.0), M7 (5.0 + 2.5) and M8 (10 + 2.5). The mixture M8 completely suppressed mycelial growth for all strains. A significant stimulation in OTA production was observed with mixtures M1 to M5 mainly at the highest a(w); whereas M6, M7 and M8 completely inhibited OTA production in all strains assayed; except M6 in A. carbonarius strain (RCP G). These results could enable a future intervention strategy to minimize OTA contamination.
Sigma-Aldrich
Propyl 4-hydroxybenzoate, ≥99%
Sigma-Aldrich
Propyl 4-hydroxybenzoate, BioXtra
Sigma-Aldrich
Propyl 4-hydroxybenzoate, tested according to Ph. Eur.
Supelco
Propylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
Propyl parahydroxybenzoate, European Pharmacopoeia (EP) Reference Standard