Merck
  • Efficacy and safety of coadministration of rosuvastatin, ezetimibe, and colestimide in heterozygous familial hypercholesterolemia.
Efficacy and safety of coadministration of rosuvastatin, ezetimibe, and colestimide in heterozygous familial hypercholesterolemia.
The American journal of cardiology (2011-11-25)
Masa-Aki Kawashiri, Atsushi Nohara, Tohru Noguchi, Hayato Tada, Chiaki Nakanishi, Mika Mori, Tetsuo Konno, Kenshi Hayashi, Noboru Fujino, Akihiro Inazu, Junji Kobayashi, Hiroshi Mabuchi, Masakazu Yamagishi
ABSTRACT
Aggressive low-density lipoprotein (LDL) cholesterol-lowering therapy is important for high-risk patients. However, sparse data exist on the impact of combined aggressive LDL cholesterol-lowering therapy in familial hypercholesterolemia (FH), particularly on side effects to changes in plasma coenzyme Q10 and proprotein convertase subtilisin/kexin type 9 levels. We enrolled 17 Japanese patients with heterozygous FH (12 men, 63.9 ± 7.4 years old) with single LDL receptor gene mutations in a prospective open randomized study. Permitted maximum doses of rosuvastatin (20 mg/day), ezetimibe (10 mg/day), and granulated colestimide (3.62 g/day) were introduced sequentially. Serum levels of LDL cholesterol decreased significantly by -66.4% (p <0.001) and 44% of participants achieved LDL cholesterol levels <100 mg/dl. There were no serious side effects or abnormal laboratory data that would have required the protocol to have been terminated except for 1 patient with myalgia. Coadministration of ezetimibe and granulated colestimide further lowered serum LDL cholesterol more than rosuvastatin alone without changing plasma coenzyme Q10 and proprotein convertase subtilisin/kexin type 9 levels. In conclusion, adequate introduction of this aggressive cholesterol-lowering regimen can improve the lipid profile of FH.
Sigma-Aldrich
(S)-(+)-Epichlorohydrin, 98%
Supelco
(±)-Epichlorohydrin, analytical standard
Sigma-Aldrich
(±)-Epichlorohydrin, purum, ≥99% (GC)