Merck
  • Sensitive turn-on fluorescent detection of tartrazine based on fluorescence resonance energy transfer.
Sensitive turn-on fluorescent detection of tartrazine based on fluorescence resonance energy transfer.
Chemical communications (Cambridge, England) (2011-11-29)
Sheng Tian Huang, Yan Shi, Nian Bing Li, Hong Qun Luo
ABSTRACT
We introduce a sensitive, rapid, label-free and general fluorescent method for the determination of tartrazine by competitive binding to reduced graphene oxide (rGO) against fluorescein, and the fluorescence recovery upon fluorescein desorption from rGO provides a quantitative readout for tartrazine, giving a detection limit of 0.53 ng mL(-1).
Sigma-Aldrich
Tartrazine, Dye content ≥85 %
Pricing and availability is not currently available.
Supelco
Tartrazine, analytical standard
Pricing and availability is not currently available.