Merck
Plasmodium species co-infection as a cause of treatment failure.
Travel medicine and infectious disease (2011-12-06)
A Smith, J Denholm, J Shortt, D Spelman
ABSTRACT
We report a case of Plasmodium falciparum and Plasmodium malariae coinfection with associated failure of clinical response to artemether + lumefantrine therapy. This case highlights the need to consider co-infection in the setting of apparent treatment failure and the impact of mixed species infection upon host dynamics and clinical presentation. Recognition of malarial co-infection is clinically important for determining appropriate therapy and preventing disease sequelae.
Sigma-Aldrich
Lumefantrine
Sigma-Aldrich
Artemether, ≥98% (HPLC)