Merck
  • Selective capture of phosphopeptides by hierarchical Ti-aluminophosphate-5 molecular sieves.
Selective capture of phosphopeptides by hierarchical Ti-aluminophosphate-5 molecular sieves.
Chemical communications (Cambridge, England) (2012-01-06)
Bo Xu, Lipeng Zhou, Fangjun Wang, Hongqiang Qin, Jun Zhu, Hanfa Zou
ABSTRACT
Hierarchical Ti-aluminophosphate-5 molecular sieves templated by glucose have been synthesized and applied as a potential adsorbent for the first time to selectively capture phosphopeptides from complex peptide mixtures prior to MALDI-TOF MS analysis.
Sigma-Aldrich
Aluminum phosphate, 99.99% trace metals basis
Sigma-Aldrich
Aluminum phosphate, reagent grade
Sigma-Aldrich
Aluminum phosphate monobasic, purum, ≥95.0% (gravimetric)
Sigma-Aldrich
Aluminum phosphate monobasic solution, ~50% in H2O