Merck
  • Pancytopenia in a patient given pyrimethamine and sulphamethoxidiazine during pregnancy.
Pancytopenia in a patient given pyrimethamine and sulphamethoxidiazine during pregnancy.
Archives of gynecology and obstetrics (1990-01-01)
A Pajor
ABSTRACT
This is a report of a pregnant woman who became pancytopenic in the second trimester following pyrimethamine administration. The patient recovered on steroid, antibiotics, transfusion, folic acid and folinic acid therapy, and the pregnancy was uneventful afterwards. The patient was delivered of a healthy term-infant.
Sigma-Aldrich
Sulfameter