Merck
  • Effects of bile-acid-binding resin (colestimide) on blood glucose and visceral fat in Japanese patients with type 2 diabetes mellitus and hypercholesterolemia: an open-label, randomized, case-control, crossover study.
Effects of bile-acid-binding resin (colestimide) on blood glucose and visceral fat in Japanese patients with type 2 diabetes mellitus and hypercholesterolemia: an open-label, randomized, case-control, crossover study.
Journal of diabetes and its complications (2012-01-14)
Tatsuya Suzuki, Kenzo Oba, Yoshimasa Igari, Kentaro Watanabe, Noriaki Matsumura, Shoko Futami-Suda, Motoshi Ouchi, Kazunari Suzuki, Ken-Ichi Sekimizu, Yoshiaki Kigawa, Hiroshi Nakano
ABSTRACT
The objective was to examine the effects of colestimide on blood glucose, visceral fat, adipocytokines, and bile acid conjugate fractions in Japanese patients. This study was an open-label, randomized, case-control, crossover study of colestimide 3 g/day in 40 Japanese patients with type 2 diabetes mellitus (T2D) and hypercholesterolemia. Patients were assigned to the colestimide group in which pravastatin and colestimide were administered orally and to the statin group in which pravastatin alone was administered orally. The principal outcome measures were serum lipid levels, fasting plasma glucose level in the early morning, hemoglobin A1c (HbA(1c)), visceral fat area (VFA), and serum 1,5-anhydroglucitol (1,5-AG) level. Serum low-density lipoprotein cholesterol levels significantly decreased from 113±38 mg/dl at baseline to 90±20 mg/dl (P=.009) at week 12 of colestimide administration. HbA(1c) significantly decreased from 7.4%±0.9% at baseline to 6.9%±0.9% (P=.001) at week 12 of colestimide administration. Serum 1,5-AG levels increased from 9.4±10.1 μg/ml to 12.4±9.5 μg/ml (P=.05) at week 12 of colestimide administration. The statin group showed no significant changes in lipids and 1,5-AG. However, ΔVFA was inversely correlated with Δcholic acid, and multivariate analysis revealed that ΔVFA was a significant explanatory variable. Colestimide holds promise not only for the treatment of hypercholesterolemia but also for the possible improvement of T2D and visceral fat obesity.
Sigma-Aldrich
1,5-Anhydro-D-sorbitol, crystalline, ≥98.0% (TLC)
Sigma-Aldrich
(±)-Epichlorohydrin, purum, ≥99% (GC)
Supelco
(±)-Epichlorohydrin, analytical standard
Sigma-Aldrich
(S)-(+)-Epichlorohydrin, 98%