Merck
  • Arsenic alters behavioral parameters and brain ectonucleotidases activities in zebrafish (Danio rerio).
Arsenic alters behavioral parameters and brain ectonucleotidases activities in zebrafish (Danio rerio).
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP (2012-01-24)
Luis Antonio Baldissarelli, Katiucia Marques Capiotti, Mauricio Reis Bogo, Gabriele Ghisleni, Carla Denise Bonan
ABSTRACT
Arsenic (As) exposure has been associated with serious chronic health risk to humans including cancer and neurological disturbances. However, there are limited studies about the mechanisms behind its toxicity. In this study, adult zebrafish were exposed to several concentrations of As (0.05, 5, and 15 mg As/L; Na(2)HAsO(4) as As(V)) during 96 h to evaluate the zebrafish locomotor activity, anxiety, and brain extracellular nucleotide hydrolysis. We showed that 5 mg/L As is able to promote significant decrease in the locomotor activity as evaluated by the number of line crossings. In addition, animals treated with 5mg/L As presented an increase in time spent in the lower zone of the tank test, suggesting an anxiogenic effect. Considering that behavioral parameters, such as anxiety and locomotion, might be modulated by the purinergic system, we also evaluated the ectonucleotidase activities in zebrafish brain after a 96-h As exposure. A significant decrease in ATP, ADP, and AMP hydrolysis was observed at 0.05, 5, and 15 mg/L when compared to control group. These findings demonstrated that As might affect behavioral parameters and the ectonucleotidase activities in zebrafish, suggesting this enzyme pathway is a target for neurotoxic effects induced by As.
Sigma-Aldrich
Sodium arsenate dibasic heptahydrate, ≥98.0%
Sigma-Aldrich
Sodium arsenate dibasic heptahydrate, ACS reagent, ≥98%