Merck
  • Rapid and sensitive determination of sulfonamide residues in milk and chicken muscle by microfluidic chip electrophoresis.
Rapid and sensitive determination of sulfonamide residues in milk and chicken muscle by microfluidic chip electrophoresis.
Journal of agricultural and food chemistry (2012-01-27)
Lili Wang, Jing Wu, Qin Wang, Chonghui He, Lei Zhou, Jing Wang, Qiaosheng Pu
ABSTRACT
A new, rapid, and sensitive method was proposed for the determination of sulfonamide residues in milk and chicken muscle samples by microchip electrophoresis with laser-induced fluorescence detection. Separation of fluorescamine-labeled sulfonamides was accomplished by using a buffer containing 5 mmol/L boric acid and 1% (w/v) polyvinyl alcohol (PVA). The pH, amount of PVA, and concentration of boric acid in the running buffer were found to have great influence on the separation. By optimizing these conditions, the separation of four sulfonamides, sulfamethazine, sulfamethoxazole, sulfaquinoxaline, and sulfanilamide, was achieved within 1 min with limits of detection (S/N = 3) of 0.2-2.3 μg/L, which are well below the maximum residue limit. The proposed method also exhibited very good repeatability; the relative standard deviations for both within-day and between-day measurements were ≤3.0%. With a simplified sample pretreatment protocol, fast determination of sulfonamides in real samples was successfully performed with standard addition recoveries of 93.3-100.8 and 82.9-92.3%, respectively.
Product Description
Supelco
Sulfanilamide melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sulfamethazine, 99.0-101.0% (on dried basis)
Supelco
Sulfanilamide, VETRANAL®, analytical standard
Supelco
Sulfamethazine, VETRANAL®, analytical standard
Sigma-Aldrich
Sulfanilamide, ≥98%
Supelco
Sulfaquinoxaline, PESTANAL®, analytical standard
Sigma-Aldrich
Sulfanilamide, puriss. p.a., ≥98% (calc. to the dried substance)