Merck
  • The role of lymphocyte proliferation tests in assessing occupational sensitization and disease.
The role of lymphocyte proliferation tests in assessing occupational sensitization and disease.
Current opinion in allergy and clinical immunology (2012-02-07)
Stella E Hines, Karin Pacheco, Lisa A Maier
ABSTRACT
Lymphocyte proliferation testing (LPT) is used in diagnosing occupationally acquired delayed-type hypersensitivity. It has been used in beryllium-health effects, and its role is expanding in metal allergy. It may find application in diagnosis of other sensitizers. Use of the beryllium LPT (BeLPT) in medical surveillance identifies beryllium sensitization at low exposure with chronic beryllium disease (CBD) that leads to physiologic impairment and need for immunosuppressive medications. New studies indicate that both beryllium exposure and genetic variation are associated with increased risk of CBD. Borderline positive BeLPTs warrant inclusion into diagnostic algorithms. Furthermore, use of LPTs to diagnose metal allergy is being proposed in diagnosis of chromium allergy and hypersensitivity to surgical implants. New occupational sensitizers continue to be identified including metalworking fluids, the sterilizing agent ortho-phthalaldehyde and the solvent para-chlorobenzotrifluoride. Use of LPT in occupational surveillance to these agents and other known sensitizers may play expanding roles. Lymphocyte proliferation testing serves a valuable role in diagnosing occupational sensitization, as demonstrated with beryllium-health effects, as cases continue to be found at low exposure levels. The use of LPTs in diagnosing contact allergy is expanding, and new applications may be identified in human and animal studies.
Sigma-Aldrich
Phthaldialdehyde, suitable for fluorescence, ≥99.0% (HPLC)
Sigma-Aldrich
Phthaldialdehyde, suitable for HPLC fluorimetric detection of amino acids, ≥99% (HPLC), powder or crystals
Sigma-Aldrich
Phthaldialdehyde, ≥97% (HPLC), powder or crystals
Sigma-Aldrich
Phthaldialdehyde Reagent, Solution Incomplete
Sigma-Aldrich
Phthaldialdehyde Reagent, Solution Complete