Merck
A triphenylene based zinc ensemble as an oxidation inhibitor.
Chemical communications (Cambridge, England) (2012-03-01)
Vandana Bhalla, Harshveer Arora, Abhimanew Dhir, Manoj Kumar
ABSTRACT
The zinc complex of a new triphenylene based receptor is evaluated for its anti-oxidant activity which is better in comparison to that of commercially available anti-oxidants.
Product Description
Triphenylene, BCR®, certified reference material
Sigma-Aldrich
Triphenylene, 98%