Merck
  • Simultaneous ingestion of dietary proteins reduces the bioavailability of galloylated catechins from green tea in humans.
Simultaneous ingestion of dietary proteins reduces the bioavailability of galloylated catechins from green tea in humans.
European journal of nutrition (2012-03-01)
Sarah Egert, Jane Tereszczuk, Silvia Wein, Manfred James Müller, Jan Frank, Gerald Rimbach, Siegfried Wolffram
ABSTRACT
To investigate the influence of dietary proteins (casein, soy protein) and skimmed milk on the plasma kinetics of green tea (GT) catechins. In a randomized cross-over design with one-week intervals, 24 healthy normal-weight women consumed a test drink containing 1.75 g GT extract with or without the addition of different proteins. Treatments were GT (control), GT with skimmed milk (GT + M), GT with caseinate (GT + CS), or GT with soy protein (GT + S). Venous blood samples were taken before and several times during a period of 4.5 h after consumption of the test drink. Plasma concentrations of catechins were analyzed by HPLC with electrochemical detection. Compared to control, consumption of GT with milk, caseinate, or soy protein significantly reduced the bioavailability (mean area under the plasma concentration-time curve) of total catechins (means ± SEM; GT + M, 87 ± 5%; GT + CS, 79 ± 5%; GT + S, 88 ± 4%), epigallocatechin gallate (GT + M, 68 ± 4%; GT + CS, 63 ± 5%; GT + S, 76 ± 5%), and epicatechin gallate (GT + M, 68 ± 5%; GT + CS, 66 ± 6%; GT + S, 77 ± 6%), while the bioavailability of non-galloylated catechins such as epigallocatechin (GT + M, 134 ± 9%; GT + CS, 118 ± 9 %; GT + S, 123 ± 8%) and epicatechin (GT + M, 125 ± 10%; GT + CS, 114 ± 11%; GT + S, 110 ± 8%) significantly increased. No significant differences in bioavailability of GT catechins were observed between the treatments GT + M, GT + CS, or GT + S. Simultaneous ingestion of dietary proteins reduces the bioavailability of galloylated catechins from GT in humans.
Product Description
Supelco
(−)-Gallocatechin, analytical standard
Sigma-Aldrich
(−)-Gallocatechin, ≥98% (HPLC)
Sigma-Aldrich
(−)-Epigallocatechin, ≥95% (HPLC), from green tea
Sigma-Aldrich
(−)-Epicatechin gallate, ≥98% (HPLC), from green tea
Supelco
(−)-Epigallocatechin, analytical standard
Epigallocatechin, primary reference standard
Epicatechin gallate, primary reference standard