Merck
  • Alpha2C-adrenoceptors play a prominent role in sympathetic constriction of porcine pulmonary arteries.
Alpha2C-adrenoceptors play a prominent role in sympathetic constriction of porcine pulmonary arteries.
Naunyn-Schmiedeberg's archives of pharmacology (2012-03-01)
Florian Jantschak, Heinz H Pertz
ABSTRACT
Enhanced pulmonary vasoconstriction in response to injuries of the central nervous system and hypoxia result in pulmonary edema due to increased sympathetic activation. This study aimed to characterize α(2)-adrenoceptor (AR)-mediated responses in porcine pulmonary arteries. α(2)-AR-mediated vasoconstriction was studied using a tissue bath protocol. α(2)-AR protein was determined by Western blotting. UK14304 (α(2)-AR agonist) elicited only a slight contraction in pulmonary arteries compared to veins. Verapamil (voltage-operated Ca(2+) channel blocker), 2-APB (store-operated Ca(2+) channel inhibitor), and P1075 (K(ATP) channel opener) induced a marked decrease of the basal tone in veins, but not in arteries. The UK14304-induced contraction in arteries was enhanced by (S)-(-)-Bay K 8644 (L-type Ca(2+) channel activator), N (ω)-nitro-L: -arginine methyl ester hydrochloride (L-NAME, eNOS inhibitor), and (S)-(-)-Bay K 8644 plus L-NAME to the same extent. Endothelium denudation failed to affect the UK14304 response. (S)-(-)-Bay K 8644 did not increase the maximal noradrenaline (non-selective α-AR agonist) or phenylephrine (α(1)-AR agonist) response. The rightward shift of the concentration-response curve to noradrenaline by prazosin (α(1)-AR antagonist) plus (S)-(-)-Bay K 8644 was smaller and non-parallel compared to that in the presence of prazosin alone. UK14304 responses were inhibited by MK912 (α(2C)-AR antagonist). Affinity of MK912 (pA(2) 9.76) and Western blotting analysis argue for an involvement of α(2C)-ARs in noradrenaline-induced contraction of pulmonary arteries. It is concluded that postjunctional α(2C)-ARs predominantly mediate contraction in porcine pulmonary arteries when the cytosolic Ca(2+) concentration is elevated. α(2C)-AR antagonists may be beneficial in the treatment of pulmonary edema.
Sigma-Aldrich
(S)-(−)-Bay K8644, ≥98% (HPLC), solid