Merck
  • Dehydrative fragmentation of 5-hydroxyalkyl-1H-tetrazoles: a mild route to alkylidenecarbenes.
Dehydrative fragmentation of 5-hydroxyalkyl-1H-tetrazoles: a mild route to alkylidenecarbenes.
Organic letters (2012-03-01)
Duncan J Wardrop, John P Komenda
ABSTRACT
The development of a mild, base-free method for the generation of alkylidenecarbenes is reported. Treatment of 5-hydroxyalkyl-1H-tetrazoles with carbodiimides generates products arising from the 1,2-rearrangement or [1,5]-C-H bond insertion of a putative alkylidenecarbene. Formation of this divalent intermediate is proposed to occur by way of a tetraazafulvene, which undergoes extrusion of 2 mol of dinitrogen. Details of this methodology, its application to the synthesis of combretastatin A-4, and an improved route to 5-hydroxyalkyl-1H-tetrazoles are described.
Sigma-Aldrich
Tetrazole solution, suitable for DNA synthesis, filtered through a 1 μm filter, ~0.45 M in acetonitrile