Merck
  • High-performance liquid chromatography with fluorescence detection and ultra-performance liquid chromatography with electrospray tandem mass spectrometry method for the determination of indoleamine neurotransmitters and their metabolites in sea lamprey plasma.
High-performance liquid chromatography with fluorescence detection and ultra-performance liquid chromatography with electrospray tandem mass spectrometry method for the determination of indoleamine neurotransmitters and their metabolites in sea lamprey plasma.
Analytica chimica acta (2012-03-13)
Huiyong Wang, Erin J Walaszczyk, Ke Li, Yu-Wen Chung-Davidson, Weiming Li
ABSTRACT
We present a comparison of two sensitive methods, HPLC with fluorescence detector (HPLC/FLD) and UPLC with electrospray tandem mass spectrometry (UPLC/MS/MS), for the determination of indoleamine neurotransmitters (NTs) and their metabolites in sea lamprey plasma samples. Liquid-liquid extraction (LLE) and solid-phase extraction (SPE) were also tested for recovery and matrix effect. The recoveries of SPE determined by HPLC/FLD and UPLC/MS/MS ranged from 75 to 123% and 78 to 105%, respectively, while the recoveries of LLE ranged from 45 to 73% and 48 to 75%, respectively. SPE combined with HPLC/FLD and UPLC/MS/MS to determine the target analytes in plasma samples were validated of the sensitivity, reproducibility, accuracy and precision. Both methods exhibited excellent linearity in the range of 0.2-50 ng mL(-1) for all analytes. The limits of detection (LOD) varied from 0.04 ng mL(-1) to 0.13 ng mL(-1) for HPLC/FLD method and 0.003 ng mL(-1) to 0.02 ng mL(-1) for UPLC/MS/MS method. The inter-day accuracy ranged from 82.5 to 127.0% for HPLC/FLD and 93.0 to 113.0% for UPLC/MS/MS. The inter-day precision ranged from 9.9 to 32.3% for HPLC/FLD and 5.4 to 13.2% for UPLC/MS/MS. These results demonstrated that the values obtained by both methods were within the satisfactory range and the UPLC/MS/MS method provided more accurate and precise measurements than HPLC/FLD method. The comparison is of great importance to determine the available detectors, considering the complexity and expensiveness versus quality parameters. These two methods were applied to the analysis of four important indoleamine neurotransmitter analytes (5-hydroxytryptamine, 5-hydroxyindole-3-acetic acid, tryptamine and melatonin) in sea lamprey plasma samples.
Sigma-Aldrich
Tryptamine hydrochloride, 99%
Sigma-Aldrich
5-Hydroxyindole, 97%
Sigma-Aldrich
Tryptamine, ≥97%