Merck
  • Properties of two molecular forms of beta-glucuronidase from the mollusc Littorina littorea L.
Properties of two molecular forms of beta-glucuronidase from the mollusc Littorina littorea L.
European journal of biochemistry (1979-01-15)
T Diez, J A Cabezas
PMID428383
ABSTRACT
The occurrence of the two molecular forms, I and II, in the beta-glucuronidase of the liver (hepatopancreas) from the marine mollusc Littorina littorea L. has been demonstrated for the first time. The two forms have been purified 355-fold and 1262-fold, respectively. Form I and II of beta-glucuronidase behave differently on DEAE-cellulose chromatography, polyacrylamide gel disc electrophoresis, isoelectric focusing (pH 5.5 and 4.2, respectively), optimum pH (4.4 and 3.4--4.1, respectively), thermal stability, Km (1.2 mM and 0.5 mM with p-nitrophenyl beta-D-glucuronide, 0.3 mM and 0.15 mM with phenolphthalein beta-D-glucuronide as substrates for form I and II, respectively) and V. Their molecular weight, estimated by gel filtration through Sephadex G-200, was about 250000 for both forms. Several subunits were separated by polyacrylamide gel electrophoresis in presence of sodium dodecyl sulphate. This beta-glucuronidase is a glycoprotein, but sialic acid(s) were not detected. The enzyme was very active on synthetic substrates and also on hexasaccharides and tetrasaccharides containing glucuronic acid residues with beta 1 leads to 3 linkages; it had practially no activity on certain glycosaminoglycans. Hg2+ and glucaro-1,4-lactone were very effective inhibitors of this enzyme; the latter by a competitive mechanism.