Merck
Speciation and release kinetics of zinc in contaminated paddy soils.
Environmental science & technology (2012-03-20)
Saengdao Khaokaew, Gautier Landrot, Rufus L Chaney, Kaumudi Pandya, Donald L Sparks
ABSTRACT
Zinc is an important nutrient for plants, but it can be toxic at high concentrations. The solubility and speciation of Zn is controlled by many factors, especially soil pH and Eh, which can vary in lowland rice culture. This study determined Zn speciation and release kinetics in Cd-Zn cocontaminated alkaline and acidified paddy soils, under various flooding periods and draining conditions, by employing synchrotron-based techniques and a stirred-flow kinetic method. Results showed almost no change in Zn speciation and release kinetics in the two soils, although the soils were subjected to different flooding periods and draining conditions. The mineral phases in which Zn is immobilized in the soil samples were constrained by linear least squares fitting (LLSF) analyses of bulk X-ray absorption fine structure (XAFS) spectra. Only two main phases were identified by LLSF, i.e., Zn-layered double hydroxides (Zn/Mg-hydrotalcite-like, and ZnAl-LDH) and Zn-phyllosilicates (Zn-kerolite). Under all soil pHs, flooding, and draining conditions, less than 22% of Zn was desorbed from the soil after a two-hour desorption experiment. The information on Zn chemistry obtained in this study will be useful in finding the best strategy to control Cd and Zn bioavailability in the Cd-Zn cocontaminated paddy soils.
Sigma-Aldrich
Triethanolamine hydrochloride, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
Triethanolamine hydrochloride, ≥99.5% (titration)
Sigma-Aldrich
Triethanolamine, puriss. p.a., ≥99% (GC)
Sigma-Aldrich
Triethanolamine, puriss., meets analytical specification of NF, ≥99% (GC)
Sigma-Aldrich
Triethanolamine, reagent grade, 98%
Sigma-Aldrich
Triethanolamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethanolamine, ≥99.0% (GC)