Merck
  • Kinetics and mechanisms of the tropospheric reactions of menthol, borneol, fenchol, camphor, and fenchone with hydroxyl radicals (OH) and chlorine atoms (Cl).
Kinetics and mechanisms of the tropospheric reactions of menthol, borneol, fenchol, camphor, and fenchone with hydroxyl radicals (OH) and chlorine atoms (Cl).
The journal of physical chemistry. A (2012-03-28)
Antonio A Ceacero-Vega, Bernabé Ballesteros, Iustinian Bejan, Ian Barnes, Elena Jiménez, José Albaladejo
ABSTRACT
Relative kinetic techniques have been used to measure the rate coefficients for the reactions of oxygenated terpenes (menthol, borneol, fenchol, camphor, and fenchone) and cyclohexanol with hydroxyl radicals (OH) and chlorine atoms (Cl) at 298 ± 2 K and atmospheric pressure. The rate coefficients obtained for the reactions of the title compounds with OH are the following (in units of 10(-11) cm(3) molecule(-1) s(-1)): (1.48 ± 0.31), (2.65 ± 0.32), (2.49 ± 0.30), (0.38 ± 0.08), (0.39 ± 0.09) for menthol, borneol, fenchol, camphor, and fenchone, respectively. For the corresponding reactions with Cl atoms the rate coefficients are as follows (in units of 10(-10) cm(3) molecule(-1) s(-1)): (3.21 ± 0.26), (3.40 ± 0.28), (2.72 ± 0.13), (2.93 ± 0.17), (1.59 ± 0.10), and (1.86 ± 0.29) for cyclohexanol, menthol, borneol, fenchol, camphor, and fenchone, respectively. The reported error is twice the standard deviation. Product studies of the reactions were performed using multipass in situ FTIR (Fourier transform infrared spectroscopy) and solid-phase microextraction (SPME) with analysis by GC-MS (gas chromatography-mass spectrometry). A detailed mechanism is proposed to justify the observed reaction products.
Sigma-Aldrich
(+)-Fenchone, purum, ≥98.0% (sum of enantiomers, GC)
Sigma-Aldrich
(+)-Borneol, 97%
Supelco
(+)-Fenchone, analytical standard
Supelco
(+)-Borneol, analytical standard
Sigma-Aldrich
L-Fenchone, ≥98%
Sigma-Aldrich
Fenchyl alcohol, ≥96%, FG
Sigma-Aldrich
(−)-Borneol, predominantly endo, 97%
Sigma-Aldrich
(1R)-(−)-Fenchone, ≥98%
Sigma-Aldrich
Isoborneol, 95%
Supelco
(−)-Borneol, analytical standard