Merck
  • [Automatic continuous monitoring of volatile organic compounds using ion mobility spectrometer array].
[Automatic continuous monitoring of volatile organic compounds using ion mobility spectrometer array].
Huan jing ke xue= Huanjing kexue (2012-04-04)
Qing-Hua Zhou, Huai-Wen Cang, Bang-Yu Ju, Lin Li, Yong-Zhai Du, Chuang Chen, Ke-Yong Hou, Jing-Hua Li, Wei-Guo Wang, Hai-Yang Li
ABSTRACT
An ion mobility spectrometer array was designed, in order to broaden the detection range of ion mobility spectrometer and improve the accuracy of compound identification. This instrument was based on the combination of ionization sources of 63Ni positive ion mode, 63Ni negative ion mode and photoionization mode with vacuum UV lamp, and it can continuously monitor the volatile organic compounds in air. With the automatic system of sampling and injection of this instrument, the positive ion of dimethyl sulfoxide and negative ion of dichloromethane were detected simultaneously. By comprehensive analysis of spectra with ion mobility spectrometer array, acrylonitrile, m-xylene and acetone were identified, which were difficult to be distinguished under the 63Ni positive ion mode. Acetone samples were determined quantitatively within four days continuously, and the results indicated that the linear range of acetone in this instrument was 2 orders of magnitude. The linear correlation coefficient R was higher than 0.995, and the relative standard deviations were controlled in the range of 4.0%-18.3%. Methacrylate leaked in simulation was monitored on-line for 24 h continuously, using the method of dynamic tracking, and the result showed the leaking time and the concentration of methacrylate directly.
Product Description
Supelco
m-Xylene, analytical standard
Supelco
m-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material