Merck
  • Chiral cyclopalladated complex promoted asymmetric synthesis of diester-substituted P,N-ligands via stepwise hydrophosphination and hydroamination reactions.
Chiral cyclopalladated complex promoted asymmetric synthesis of diester-substituted P,N-ligands via stepwise hydrophosphination and hydroamination reactions.
Dalton transactions (Cambridge, England : 2003) (2012-04-05)
Ke Chen, Sumod A Pullarkat, Mengtao Ma, Yongxin Li, Pak-Hing Leung
ABSTRACT
A series of enantiomerically pure 1,2-diester substituted P,N-ligands incorporating two chiral carbons in the backbone were generated in high yields and high stereoselectivity from acetylenedicarboxylate via initial hydrophosphination using diphenylphosphine followed by hydroamination with various primary and secondary amines. The reactions were activated and stereochemically controlled by the organopalladium complex containing ortho-palladated (S)-(1-(dimethylamino)ethyl)naphthalene under mild conditions. The absolute stereochemistry and the coordination chemistry of P,N-products were determined by the single crystal X-ray diffraction analysis. All the chiral P,N-ligands could be liberated from the palladium template without loss of optical purity. Subsequent recomplexation to selected chiral palladium centers confirmed the optical purity of the new functionalized chiral P,N-ligands.
Product Description
Supelco
Maleic acid, qNMR standard, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Sodium maleate dibasic, purum p.a., anhydrous, ≥98.0% (NT)
Sigma-Aldrich
Maleic acid, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Maleic acid sodium salt, ~99%