Merck
  • Coloring-decoloring behavior of amphiphilic fluoroalkyl end-capped N-(1,1-dimethyl-3-oxobutyl)acrylamide--acryloylmorpholine cooligomer/fluorescein nanocomposites in protic and aprotic solvents.
Coloring-decoloring behavior of amphiphilic fluoroalkyl end-capped N-(1,1-dimethyl-3-oxobutyl)acrylamide--acryloylmorpholine cooligomer/fluorescein nanocomposites in protic and aprotic solvents.
Journal of colloid and interface science (2012-04-10)
Hideo Sawada, Shunsuke Izumi, Kazuo Sasazawa, Masato Yoshida
ABSTRACT
Amphiphilic fluoroalkyl end-capped N-(1,1-dimethyl-3-oxobutyl)acrylamide-acryloylmorpholine cooligomer/fluorescein nanocomposites afforded brilliant yellow-colored solutions in not only protic solvents such as methanol and ethanol but also protic-like solvents such as dichloromethane and 1,2-dichloroethane, respectively. However, the corresponding non-fluorinated cooligomer/fluorescein composites and parent fluorescein gave the colorless solutions under similar conditions. On the other hand, unexpectedly, such brilliant yellow-colored solutions provided by these fluorinated nanocomposites completely disappeared in aprotic solvents such as N,N-dimethylformamide, dimethyl sulfoxide, and tetrahydrofuran. Thus, these fluorinated fluorescein nanocomposites can exhibit a coloring-decoloring behavior through solvatochromic response.
Product Description
Supelco
1,2–Dichloroethane solution, certified reference material, 200 μg/mL in methanol
Supelco
1,2-Dichloroethane, analytical standard
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, 99.8%
Sigma-Aldrich
1,2-Dichloroethane, ACS reagent, ≥99.0%