Merck
  • Reactivity toward oxygen radicals and antioxidant action of thiol compounds.
Reactivity toward oxygen radicals and antioxidant action of thiol compounds.
BioFactors (Oxford, England) (2012-04-11)
Mizuki Takashima, Mototada Shichiri, Yoshihisa Hagihara, Yasukazu Yoshida, Etsuo Niki
ABSTRACT
Thiol compounds exert diverse functions in the defense network against oxidative stress in vivo. Above all, the role of glutathione in the enzymatic removal of hydrogen peroxide and lipid hydroperoxides has been well established. The scavenging of reactive free radicals is one of the many functions. In this study, the reactivities of several thiol compounds toward oxygen- and nitrogen-centered radicals were measured from their reaction with galvinoxyl and 1,1-diphenyl-2-picrylhydrazyl (DPPH) radicals and also from their sparing effects on the decay of fluorescein, pyrogallol red, and BODIPY induced by peroxyl radicals. Furthermore, the antioxidant capacity against lipid peroxidation was assessed in the oxidation of methyl linoleate induced by free radicals in micelle systems. Cysteine, homocysteine, and glutathione exhibited considerable reactivity toward galvinoxyl, DPPH, and peroxyl radicals in this order but methionine did not. Bovine serum albumin (BSA) was less reactive toward these radicals than cysteine on molar base. Cysteine, homocysteine, and glutathione suppressed the oxidation of methyl linoleate in micelle systems, but methionine did not. The reactivity toward free radicals and antioxidant capacity of these thiol compounds were less than that of ascorbic acid, but higher than that of uric acid.
Sigma-Aldrich
Galvinoxyl, free radical
Pricing and availability is not currently available.