Merck
  • Chemical diversity of essential oils from Asteriscus graveolens (Forssk.) less.: identification of cis-8-Acetoxychrysanthenyl acetate as a new natural component.
Chemical diversity of essential oils from Asteriscus graveolens (Forssk.) less.: identification of cis-8-Acetoxychrysanthenyl acetate as a new natural component.
Chemistry & biodiversity (2012-04-12)
Gregory Cristofari, Mohamed Znini, Lhou Majidi, Hamid Mazouz, Pierre Tomi, Jean Costa, Julien Paolini
ABSTRACT
Asteriscus graveolens is an endemic medicinal plant mainly distributed in south-western Algeria and south-eastern Morocco. The essential oils of leaves, stems, and flowers of A. graveolens had been studied by GC, GC/MS, and (13) C-NMR. The spectral data of two nerolidol derivatives, 6-oxo- and 6-hydroxycyclonerolidol, were reassigned by 1D- and 2D-NMR spectroscopy. These compounds can be considered as chemical markers of this genus. The structure of a monoterpenic diester with a chrysanthenane skeleton, i.e., cis-8-acetoxychrysanthenyl acetate, was determined for the first time on the basis of GC/MS, and 1D- and 2D-NMR. The stem and leaf oils were characterized by high content of oxygenated sesquiterpenes with 6-oxo- and 6-hydroxycyclonerolidol as major components, and the flower essential oils were dominated by the new monoterpenic compound cis-8-acetoxychrysanthenyl acetate.
Product Description
Supelco
trans-Nerolidol, analytical standard
Sigma-Aldrich
Nerolidol, 98%, Mixture of cis and trans
Sigma-Aldrich
Nerolidol, mixture of cis and trans, ≥97%, stabilized, FG