Merck
  • [Synthesis of 2'-aminometylmorpholino nucleoside analogues containing 4'-carboxymethyl lynker group].
[Synthesis of 2'-aminometylmorpholino nucleoside analogues containing 4'-carboxymethyl lynker group].
Bioorganicheskaia khimiia (2012-04-14)
M F Kasakin, T V Abramova, V N Sil'nikov
ABSTRACT
A simple and effective method for the synthesis of 2'-aminomethylmorpholino-4'-carboxymethyl nucleoside analogues and Boc-modified derivatives as synthons for peptide synthesis was developed.
Sigma-Aldrich
Morpholine, ReagentPlus®, ≥99%
Sigma-Aldrich
Morpholine, ACS reagent, ≥99.0%
Supelco
Morpholine, analytical standard
Sigma-Aldrich
Morpholine, purified by redistillation, ≥99.5%