Merck
  • Effect of abamectin exposure on semen parameters indicative of reduced sperm maturity: a study on farmworkers in Antalya (Turkey).
Effect of abamectin exposure on semen parameters indicative of reduced sperm maturity: a study on farmworkers in Antalya (Turkey).
Andrologia (2012-04-26)
C Celik-Ozenci, A Tasatargil, M Tekcan, L Sati, E Gungor, M Isbir, M F Usta, M E Akar, F Erler
ABSTRACT
Environmental exposure to pesticides may cause serious health risks including fertility and reproductive function. The aim of this study was to highlight whether there is a relationship between exposure to abamectin and male fertility parameters of farmworkers. Twenty male farmworkers who were using abamectin and 20 men not exposed to pesticides were recruited as experimental and control groups, respectively. Semen analysis, molecular markers of sperm maturity and serum reproductive hormone levels were evaluated. In experimental group, high plasma abamectin levels were detected. These men have decreased sperm motility. Moreover, diminished molecular markers of sperm maturity, such as decreased hyaluronic acid (HA) binding of sperm, increased numbers of aniline blue positive sperm and increased percentage of creatine kinase (CK) positive sperm, were observed in abamectin-exposed men. Their serum testosterone, LH and FSH levels did not change significantly. We conclude that exposure to abamectin may impair male fertility by effecting semen quality.
Product Description
Supelco
Abamectin, PESTANAL®, analytical standard