Merck
Hybrid multiferroic nanostructure with magnetic-dielectric coupling.
Nano letters (2012-05-02)
T N Narayanan, B P Mandal, A K Tyagi, A Kumarasiri, Xaiobo Zhan, Myung Gwan Hahm, M R Anantharaman, G Lawes, P M Ajayan
ABSTRACT
The development of methods to economically synthesize single wire structured multiferroic systems with room temperature spin-charge coupling is expected to be important for building next-generation multifunctional devices with ultralow power consumption. We demonstrate the fabrication of a single nanowire multiferroic system, a new geometry, exhibiting room temperature magnetodielectric coupling. A coaxial nanotube/nanowire heterostructure of barium titanate (BaTiO(3), BTO) and cobalt (Co) has been synthesized using a template-assisted method. Room temperature ferromagnetism and ferroelectricity were exhibited by this coaxial system, indicating the coexistence of more than one ferroic interaction in this composite system.
Sigma-Aldrich
Barium titanate(IV), nanopowder (cubic crystalline phase), <100 nm particle size (BET), ≥99% trace metals basis
Sigma-Aldrich
Barium titanate(IV), powder, <3 μm, 99%
Sigma-Aldrich
Barium titanate(IV), powder, <2 μm, 99.5% trace metals basis