Merck
  • Fluorescence immunosensor based on p-acid-encapsulated silica nanoparticles for tumor marker detection.
Fluorescence immunosensor based on p-acid-encapsulated silica nanoparticles for tumor marker detection.
The Analyst (2012-05-05)
Mei Yan, Shenguang Ge, Weiqiang Gao, Chengchao Chu, Jinghua Yu, Xianrang Song
ABSTRACT
A novel, simple and highly sensitive amplified fluorescence label-free immunosensor by using p-acid-encapsulated silica nanomaterials has been developed for the first time. 4,4'-(2,5-Dimethoxy-1,4-phenylene)bis(ethyne-2,1-diyl)dibenzoic acid (p-acid) and p-acid-encapsulated silica were prepared, and characterized by Fourier transform infrared spectroscopy, nuclear magnetic resonance, ultraviolet visible spectroscopy (UV-vis) and fluorescence spectroscopy. In layer-by-layer self-assembling processes using (3-aminopropyl)triethoxysilane, p-acid@SiO(2) was assembled on the glass substrate. Antibody was immobilized on the surface of p-acid@SiO(2) with N,N'-carbonyldiimidazole. The functional nanomaterials present good analytical properties with a calibration range of 0.1-100 ng mL(-1), and allow the detection of carcinoembryonic antigen (CEA) at a concentration as low as 0.04 ng mL(-1). What is important is that the as-synthesized p-acid@SiO(2) nanomaterials could be further extended for the detection of other biomarkers or biocompounds.
Sigma-Aldrich
CDI, ≥97.0% (T)
Sigma-Aldrich
CDI, reagent grade