Merck
  • Gas chromatography-mass spectrometry of hexafluoroacetone derivatives: First time utilization of a gaseous phase derivatizing agent for analysis of extraterrestrial amino acids.
Gas chromatography-mass spectrometry of hexafluoroacetone derivatives: First time utilization of a gaseous phase derivatizing agent for analysis of extraterrestrial amino acids.
Journal of chromatography. A (2012-05-29)
C Geffroy-Rodier, A Buch, R Sternberg, S Papot
ABSTRACT
Within the perspective of the current and next space missions to Mars (MSL 2011 and Exomars 2016-2018), the detection and enantioselective separation of building blocks such as the amino acids are important subjects which are becoming fundamental for the search for traces of life on the surface and subsurface of Mars. In this work, we have developed and optimized a method adapted to space experimentation to derivatize and analyze amino acids, using hexafluoroacetone as the derivatizing agent. The temperature, duration of the derivative transfer to the analyser, and chromatographic separation parameters have been optimized to meet the instrument design constraints imposed on devices for extraterrestrial experiments. The work presented in this rationale has established that hexafluoroacetone, in addition to its intrinsic qualities, such as the production of light-weight derivatives (no racemization) and great resistance to the drastic operating conditions, has indeed facilitated simple and fast derivatization that appears to be suitable for in situ analysis in space. By using hexafluoroacetone as the derivatizing agent, we successfully identified, 21 amino acids including 12 of the 20 proteinic amino acids without stirring or extraction steps. Ten of these derivatized amino acids were enantioselectively separated. The precision and accuracy measurements for the D/L ratio showed that the proposed method was also suitable for the determination of both enantioselective forms of most of the tested amino acids. The limits of detection obtained were lower than the ppb level of organic molecules detected in Martian meteorites.
Sigma-Aldrich
Hexafluoroacetone trihydrate, 98%