Merck
  • Effects of the tropical ginger compound,1'-acetoxychavicol acetate, against tumor promotion in K5.Stat3C transgenic mice.
Effects of the tropical ginger compound,1'-acetoxychavicol acetate, against tumor promotion in K5.Stat3C transgenic mice.
Journal of experimental & clinical cancer research : CR (2012-06-19)
Vinita Batra, Zanobia Syed, Jennifer N Gill, Malari A Coburn, Patrick Adegboyega, John DiGiovanni, J Michael Mathis, Runhua Shi, John L Clifford, Heather E Kleiner-Hancock
ABSTRACT
The purpose of the current study was to determine whether a tropical ginger derived compound 1'-acetoxychavicol acetate (ACA), suppresses skin tumor promotion in K5.Stat3C mice. In a two-week study in which wild-type (WT) and K5.Stat3C mice were co-treated with either vehicle, ACA, galanga extract, or fluocinolone acetonide (FA) and tetradecanoyl phorbol acetate (TPA), only the galanga extract and FA suppressed TPA-induced skin hyperproliferation and wet weight. None of these agents were effective at suppressing p-Tyr705Stat3 expression. However, ACA and FA showed promising inhibitory effects against skin tumorigenesis in K5.Stat3C mice. ACA also suppressed phospho-p65 NF-κB activation, suggesting a potential mechanism for its action.
Product Description
Fluocinolone acetonide, European Pharmacopoeia (EP) Reference Standard
Supelco
Fluocinolone acetonide, analytical standard