Merck
  • Reduction of antibiotics using microorganisms containing glutathione S-transferases under immobilized conditions.
Reduction of antibiotics using microorganisms containing glutathione S-transferases under immobilized conditions.
Environmental toxicology and pharmacology (2012-06-19)
Hyeyoung Park
ABSTRACT
The degradation of several antibiotics (tetracycline, sulfathiazole, ampicillin) was performed with immobilized bacterial cells containing the glutathione S-transferases (GSTs). Antibiotics in animal feed contaminated wastewater usually inhibit the growth of microorganisms that treat the wastewater, so a bio-friendly treatment method is required. Therefore, we have shown that the inhibitory effects of antibiotics on bacteria were reduced by microorganisms containing detoxifying enzyme GSTs by using a cell immobilizing method in a bioreactor. The initial concentrations of tetracycline, sulfathiazole and ampicillin were 100mg/L, 100mg/L and 50mg/L respectively, which are typical of the range detected in pig feed in Korea. In the results, we observed the removal efficiency of tetracycline to be almost 70% with Staphylococcus epidermidis in the bioreactor, suggesting that this method of antibiotic removal is worthy of further study.
Product Description
Supelco
Sulfathiazole, VETRANAL®, analytical standard
Supelco
Sulfathiazole