Merck
  • Interaction of nucleic acids with Coomassie Blue G-250 in the Bradford assay.
Interaction of nucleic acids with Coomassie Blue G-250 in the Bradford assay.
Analytical biochemistry (2012-06-30)
Broc R Wenrich, Toni A Trumbo
ABSTRACT
The Bradford assay has been used reliably for decades to quantify protein in solution. The analyte is incubated in acidic solution of Coomassie Blue G-250 dye, during which reversible ionic and nonionic binding interactions form. Bradford assay color yields were determined for salmon, bovine, shrimp, and kiwi fruit genomic DNA; baker's yeast RNA; bovine serum albumin (BSA); and hen egg lysozyme. Pure DNA and RNA bound the dye, with color yields of 0.0017 mg⁻¹ cm⁻¹ and 0.0018 mg⁻¹ cm⁻¹, respectively. The nucleic acid-Coomassie Blue response was significant, at roughly 9% of that for BSA and 18% of that for lysozyme.
Sigma-Aldrich
Brilliant Blue G solution, Concentrate
Sigma-Aldrich
Brilliant Blue G, pure
Sigma-Aldrich
Brilliant Blue G, 250, suitable for microscopy