Merck
  • Comparison of micrometeorological and two-film estimates of air-water gas exchange for alpha-hexachlorocyclohexane in the Canadian archipelago.
Comparison of micrometeorological and two-film estimates of air-water gas exchange for alpha-hexachlorocyclohexane in the Canadian archipelago.
Environmental science and pollution research international (2012-07-07)
Fiona Wong, Liisa M Jantunen, Tim Papakyriakou, Ralf M Staebler, Gary A Stern, Terry F Bidleman
ABSTRACT
The air-sea gas exchange of alpha-hexachlorocyclohexane (α-HCH) in the Canadian Arctic was estimated using a micrometeorological approach and the commonly used Whitman two-film model. Concurrent shipboard measurements of α-HCH in air at two heights (1 and 15 m) and in surface seawater were conducted during the Circumpolar Flaw Lead study in 2008. Sampling was carried out during eight events in the early summer time when open water was encountered. The micrometeorological technique employed the vertical gradient in air concentration and the wind speed to estimate the flux; results were corrected for atmospheric stability using the Monin-Obukhov stability parameter. The Whitman two-film model used the concentrations of α-HCH in surface seawater, in bulk air at 1 and 15 m above the surface, and the Henry's law constant adjusted for temperature and salinity to derive the flux. Both approaches showed that the overall net flux of α-HCH was from water to air. Mean fluxes calculated using the micrometeorological technique ranged from -3.5 to 18 ng m(-2) day(-1) (mean 7.4), compared to 3.5 to 14 ng m(-2) day(-1) (mean 7.5) using the Whitman two-film model. Flux estimates for individual events agreed in direction and within a factor of two in magnitude for six of eight events. For two events, fluxes estimated by micrometeorology were zero or negative, while fluxes estimated with the two-film model were positive, and the reasons for these discrepancies are unclear. Improvements are needed to shorten air sampling times to ensure that stationarity of meteorological conditions is not compromised over the measurement periods. The micrometeorological technique could be particularly useful to estimate fluxes of organic chemicals over water in situations where no water samples are available.
Product Description
Supelco
HCH, PESTANAL®, analytical standard, mixture of isomers (α:β:γ:δ=1:1:1:1)
Supelco
α-HCH, PESTANAL®, analytical standard