Merck
Enzyme or electrode?
Structure (London, England : 1993) (2012-07-10)
Oliver Einsle
ABSTRACT
The process of dissimilatory metal reduction shapes our environment on a global scale by using minerals as terminal acceptors in a biological electron transport chain employed by bacteria under anaerobic conditions. In this issue of Structure, Edwards et al. present the structure of an extracellular undecaheme cytochrome involved in the step of electron transfer to metal oxides.
Sigma-Aldrich
Nitrilotriacetic acid, ACS reagent, ≥99.0%
Supelco
Iron(III) citrate tribasic monohydrate, 18-20% Fe basis (T)
Sigma-Aldrich
Iron(III) citrate, technical grade
Sigma-Aldrich
Nitrilotriacetic acid, Sigma Grade, ≥99%
Sigma-Aldrich
Nitrilotriacetic acid, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ferric citrate, BioReagent, suitable for cell culture